Nikotinsyra (Niacin)

Leverantör: RESEARCH ORGANICS
Varning

Synonyms: 3-Pyridinecarboxylic acid, Pyridine-3-carboxylic acid, 3-Picolinic acid, Niacin, Nicotinic acid, Vitamin B3

RES11046-A101X RES11046-A102X
RESERES11046-A101XEA 0 SEK
RESERES11046-A101X RESERES11046-A102X
Nikotinsyra (Niacin)
Nikotinsyra (Niacin)
Formel: C₆H₅NO₂
MW: 123,11 g/mol
Kokpunkt: 258 °C (1013 hPa)
Smältpunkt: 142 °C
Densitet: 1,473 g/cm³ (20 °C)
Flampunkt: 193 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006391
CAS nummer: 59-67-6
EINECS: 200-441-0
Merck Index: 13,06552

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR