Isonikotinoylhydrazid, Supelco®
Leverantör: Merck
Varning
Synonyms: 4-Picolinic acid hydrazide, 4-Pyridinecarboxylic acid hydrazide, Isonicotinic hydrazide, Pyridine-4-carboxylic acid hydrazide, Isonicotinic acid hydrazide
I3377-250G I3377-50G I3377-5G
I3377-250GEA 1830 SEK
I3377-250G I3377-50G I3377-5G
Isonikotinoylhydrazid, Supelco®
Isonikotinoylhydrazid
Formel: C₆H₇N₃O
MW: 137,14 g/mol
Smältpunkt: 169…172 °C
Densitet: 1,42 g/cm³ (20 °C)
Flampunkt: >250 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006426
CAS nummer: 54-85-3
EINECS: 200-214-6
Merck Index: 13,05205
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4d8d457734ee774427c90f9c93b5a333d84481d&vid=f4d8d457734ee774427c90f9c93b5a333d84481d&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cb74e9609cb47084b41065561f23105238fafae&vid=9cb74e9609cb47084b41065561f23105238fafae&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=007effce2e20ff4f0c11b07dd55021ebc2891210&vid=007effce2e20ff4f0c11b07dd55021ebc2891210&prid=web&clid=SAPDAM