Isonicotinonitrile 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Pyridinecarbonitrile, Pyridine-4-carbonitrile, 4-Cyanopyridine
110871000 110875000 110870025 110870250
ACRO110871000EA 545 SEK
ACRO110871000 ACRO110875000 ACRO110870025 ACRO110870250
Isonicotinonitrile 98%
Isonicotinonitrile
Formel: C₆H₄N₂
MW: 104,11 g/mol
Kokpunkt: 196 °C (760 mmHg)
Smältpunkt: 77…81 °C
Flampunkt: 88 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006417
CAS nummer: 100-48-1
EINECS: 202-856-2
Order Now
Specifikation testresultat
Appearance White to yellow to beige to light brown Powder or crystals or flakes
Infrared spectrum Conforms
Melting point 76 to 80 °C
GC ≥97.5 %