3-Amino-2,6-dichloropyridine

Leverantör: Thermo Scientific

Synonyms: 2,6-Dichloro-3-pyridinamine, 2,6-Dichloropyridin-3-amine, 2,6-Dichloro-3-pyridylamine

RF05052DA RF05052EA
MAYBRF05052DAEA 0 SEK
MAYBRF05052DA MAYBRF05052EA
3-Amino-2,6-dichloropyridine
3-Amino-2,6-dichloropyridine
Formel: C₅H₄Cl₂N₂
MW: 163,01 g/mol
Kokpunkt: 305 °C (1013 hPa)
Smältpunkt: 119 °C
Densitet: 1,497 g/cm³ (20 °C)
MDL Number: MFCD00023417
CAS nummer: 62476-56-6
EINECS: 263-559-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR