2-Chloroisonicotinamide

Leverantör: Thermo Scientific

SPB06910DA SPB06910EA
MAYBSPB06910DAEA 0 SEK
MAYBSPB06910DA MAYBSPB06910EA
2-Chloroisonicotinamide
2-Chloroisonicotinamide
Formel: C₆H₅ClN₂O
MW: 156.57 g/mol
MDL Number: MFCD00221401
CAS nummer: 100859-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR