2-Chloronicotinonitrile 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Chloro-3-cyanopyridine, 2-Chloropyridine-3-carbonitrile
352080010 352080050 352080250
ACRO352080010EA 445 SEK
ACRO352080010 ACRO352080050 ACRO352080250
2-Chloronicotinonitrile 98%
2-Chloronicotinonitrile
Formel: C₆H₃ClN₂
MW: 138,56 g/mol
Kokpunkt: 112 °C (1 mmHg)
Smältpunkt: 107…109 °C
Storage Temperature: Omgivande
MDL Number: MFCD00014628
CAS nummer: 6602-54-6
EINECS: 229-550-1
UN: 3439
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 105 to 110 °C
GC ≥97.5 %