2-Amino-5-bromopyridine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 5-Bromo-2-pyridylamine, 5-Bromopyridin-2-amine

A1587-250G A1587-25G
TCIAA1587-250GEA 2375 SEK
TCIAA1587-250G TCIAA1587-25G
2-Amino-5-bromopyridine ≥98.0% (by GC, titration analysis)
2-Amino-5-bromopyridine
Formel: C₅H₅BrN₂
MW: 173,01 g/mol
Kokpunkt: 244 °C (1013 hPa)
Smältpunkt: 136…139 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006323
CAS nummer: 1072-97-5
EINECS: 214-019-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5818ca204b0255000f252738145346b8cca104e&vid=b5818ca204b0255000f252738145346b8cca104e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 136.0 to 141.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR