2-Amino-3,5-dibromopyridine

Leverantör: Thermo Scientific
Varning

Synonyms: 3,5-Dibromo-2-pyridylamine, 3,5-Dibromopyridin-2-amine

BTB07310EA
MAYBBTB07310EAEA 0 SEK
MAYBBTB07310EA
2-Amino-3,5-dibromopyridine
2-Amino-3,5-dibromopyridine
Formel: C₅H₄Br₂N₂
MW: 251,91 g/mol
Kokpunkt: 254 °C (1013 hPa)
Smältpunkt: 104 °C
MDL Number: MFCD00038041
CAS nummer: 35486-42-1
EINECS: 252-590-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR