Nikotinamid-adenin-dinukleotidlösning (oxiderad form), Millipore®
Leverantör: Merck
Synonyms: Nadide, NAD+, Nicotinamide adenine dinucleotide, oxidized
M-Clarity™ quality level = MQ200
1.24542.0001 1.24542.0005 1.24542.0025
1.24542.0001EA 616 SEK
1.24542.0001 1.24542.0005 1.24542.0025
Nikotinamid-adenin-dinukleotidlösning (oxiderad form), Millipore®
Nikotinamid-adenin-dinukleotidlösning (oxiderad form)
Formel: C₂₁H₂₇N₇O₁₄P₂
MW: 663,43 g/mol
Smältpunkt: 140 to 142 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00036253
CAS nummer: 53-84-9
EINECS: 200-184-4
Merck Index: 13,06370