Nikotinamid-adenin-dinukleotidlösning (oxiderad form), Millipore®
Leverantör: Merck Millipore Calbiochem
Synonyms: Nadide, NAD+, Nicotinamide adenine dinucleotide, oxidized
481911-10 481911-5
481911-10EA 11875 SEK
481911-10 481911-5
Nikotinamid-adenin-dinukleotidlösning (oxiderad form), Millipore®
Nikotinamid-adenin-dinukleotidlösning (oxiderad form)
Formel: C₂₁H₂₇N₇O₁₄P₂
MW: 663,43 g/mol
Smältpunkt: 140 to 142 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00036253
CAS nummer: 53-84-9
EINECS: 200-184-4
Merck Index: 13,06370