Adenin ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine
A14906.14 A14906.18 A14906.30 A14906.36 A14906.A7
A14906.14EA 1350 SEK
A14906.14 A14906.18 A14906.30 A14906.36 A14906.A7
Adenin ≥99%
Adenin
Purity: 99% CAS No.: 73-24-5 Molecular Formula: C5H5N5 Formula Weight: 135.13 Odour: Odourless Melting Point: 360° dec. Beilstein Reference: 5777 Merck Reference: 14,152 UN No.: 2811 MDL No.: MFCD00041790
Formel: C₅H₅N₅
MW: 135,13 g/mol
Smältpunkt: >360 °C (dec.)
Densitet: 0,99741 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00041790
CAS nummer: 73-24-5
EINECS: 200-796-1
UN: 2811
ADR: 6.1,III
Merck Index: 14,152