2-Aminopurine

Leverantör: Thermo Scientific
Varning

Synonyms: Purin-2-amine, 7H-Purin-2-amine

AC27656.250
MAYBAC27656.250EA 0 SEK
MAYBAC27656.250
2-Aminopurine
2-Aminopurine
Formel: C₅H₅N₅
MW: 135,13 g/mol
Smältpunkt: 277…278 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005566
CAS nummer: 452-06-2
EINECS: 207-197-4

Order Now

Specifikation testresultat

Appearance Light beige Powder
Infrared spectrum Conforms
Melting point 280°C to 284°C
Assay ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR