Etidiumbromid 98 Torrvikt
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 3,8-Diamino-5-etyl-6-fenylfenantridiniumbromid
L07482.03 L07482.06
L07482.03EA 985 SEK
L07482.03 L07482.06
Etidiumbromid 98 Torrvikt
Etidiumbromid
Purity: 98% CAS No.: 1239-45-8 Molecular Formula: C21H20BrN3 Formula Weight: 394.32 Odour: Weak Melting Point: ca 261° dec. Beilstein Reference: 3642536 Merck Reference: 14,4731 UN No.: 2811 MDL No.: MFCD00011724
Formel: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
Kokpunkt: 72 °C (0.1 mmHg)
Smältpunkt: ∼ 261 °C (dec.)
Densitet: 1.01 g/cm³ (20 °C)
Flampunkt: <100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011724
CAS nummer: 1239-45-8
EINECS: 214-984-6
UN: 2811
ADR: 6.1 I
Merck Index: 14,4731