Natriumdehydracetat ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: Natrium-1-(3,4-dihydro-6-metyl-2,4-dioxo-2H-pyran-3-yliden)etanolat
B21060.0E B21060.22 B21060.36
B21060.0EEA 11225 SEK
B21060.0E B21060.22 B21060.36
Natriumdehydracetat ≥97%
Natriumdehydracetat
Purity: 97% CAS No.: 4418-26-2 Molecular Formula: C8H7NaO4 Formula Weight: 190.13 pH: 8.8-9.3 Melting Point: ca 295° dec. MDL No.: MFCD00040583
Formel: C₈H₇NaO₄
MW: 190,13 g/mol
Smältpunkt: ∼ 295 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00040583
CAS nummer: 4418-26-2
EINECS: 224-580-1