Dehydracetsyra ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3-Acetyl-6-metyl-2H-pyran-2,4(3H)-dion
A17827.0B A17827.18 A17827.30
A17827.0BEA 3950 SEK
A17827.0B A17827.18 A17827.30
Dehydracetsyra ≥98%
Dehydracetsyra
Purity: 98% CAS No.: 520-45-6 Molecular Formula: C8H8O4 Formula Weight: 168.15 Flash Point: 157°(314°F) Melting Point: 109-113° Boiling Point: 269-270° Beilstein Reference: 6129 Merck Reference: 14,2865 MDL No.: MFCD00066709
Formel: C₈H₈O₄
Kokpunkt: 269…270 °C
Smältpunkt: 109…113 °C
Flampunkt: 157 °C (314 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00066709
CAS nummer: 520-45-6
EINECS: 208-293-9
Merck Index: 14,2865