L (+)-Askorbinsyra ≥98%
Leverantör: Thermo Fisher Scientific
11188.22 11188.36 11188.A3
11188.22EA 805 SEK
11188.22 11188.36 11188.A3
L (+)-Askorbinsyra ≥98%
L (+)-Askorbinsyra
Purity: 98+% CAS No.: 50-81-7 Molecular Formula: C6H8O6 Formula Weight: 176.12 Physical Form: Powder Density: 1.65 Melting Point: ca 191° dec. Beilstein Reference: 84272 Merck Reference: 14,830 MDL No.: MFCD00064328
Formel: C₆H₈O₆
Kokpunkt: 553 °C (1013 hPa)
Smältpunkt: ∼ 191 °C (dec.)
Densitet: 1,65
Storage Temperature: Omgivande
MDL Number: MFCD00064328
CAS nummer: 50-81-7
EINECS: 200-066-2
REACH: exempt
Merck Index: 14,830
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c03499908e269560d680b0d7c4651f97a367aa0&vid=73a720d6900e713a98164022273a8afa6114ad7b&prid=web&clid=SAPDAM