Kobolt(II)sulfat, heptahydrat 99+%, extra pure
Leverantör: Thermo Scientific
Fara
Synonyms: Cobalt sulphate heptahydrate, Cobaltous sulphate heptahydrate
213101000 213105000 213100025
ACRO213101000EA 1400 SEK
ACRO213101000 ACRO213105000 ACRO213100025
Kobolt(II)sulfat, heptahydrat 99+%, extra pure
Kobolt(II)sulfat, heptahydrat
Formel: CoSO₄·7H₂O
MW: 281,1 g/mol
Smältpunkt: 98 °C
Densitet: 2,03 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149658
CAS nummer: 10026-24-1
EINECS: 233-334-2
UN: 3077
ADR: 9,III
Merck Index: 12,02510
Order Now
Specifikation testresultat
Appearance Red-brown Adhering crystalline powder or crystals
Titration Complexometric ≥99.0 %
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm
Nickel (Ni) ≤500 ppm