Järn(II)sulfid, tech.
Leverantör: Thermo Scientific
Synonyms: Ferrous sulphide
A15569.0B A15569.0I A15569.30
A15569.0BEA 2600 SEK
A15569.0B A15569.0I A15569.30
Järn(II)sulfid, tech.
Järn(II)sulfid
CAS No.: 1317-37-9 Molecular Formula: FeS Formula Weight: 87.91 Odour: Odourless Melting Point: 1195° Merck Reference: 14,4058 MDL No.: MFCD00011013
Formel: FeS
MW: 87,91 g/mol
Smältpunkt: 1195 °C
Densitet: 4,74
Storage Temperature: Omgivande
MDL Number: MFCD00011013
CAS nummer: 1317-37-9
EINECS: 215-268-6
UN: 3077
ADR: 9,III
Merck Index: 14,4058