Ruthenium(IV) oxide, vattenfri 99.9%
Leverantör: Thermo Fisher Scientific
Fara
A10816.14 A10816.03 A10816.06
A10816.14EA 49800 SEK
A10816.14 A10816.03 A10816.06
Ruthenium(IV) oxide, vattenfri 99.9%
Ruthenium(IV) oxide
Purity: 99.9% CAS No.: 12036-10-1 Molecular Formula: RuO2 Formula Weight: 133.07 Odour: Odourless Melting Point: 1200° subl. UN No.: 1479 MDL No.: MFCD00011210
Formel: O₂Ru
Smältpunkt: 1200 °C (subl.)
Densitet: 6,97
Storage Temperature: Omgivande
MDL Number: MFCD00011210
CAS nummer: 12036-10-1
EINECS: 234-840-6
UN: 1479
ADR: 5.1,II