Ruthenium(IV) oxide, vattenfri ≥99.95% Ru ≥ 75.2%, Premion®
Leverantör: Thermo Fisher Scientific
|
Fara
|
Synonyms:
Ruthenium dioxide, Ruthenium tetroxide
11804.02
11804.04
11804.09
11804.02EA
1200
SEK
11804.02
11804.04
11804.09
Ruthenium(IV) oxide, vattenfri ≥99.95% Ru ≥ 75.2%, Premion®
Ruthenium(IV) oxide
Purity: 99.95%
CAS No.: 12036-10-1
Molecular Formula: O2Ru
Formula Weight: 133.07
Physical Form: Powder
Density: 6.97
Appearance: Black
Odour: Odourless
Melting Point: 1200° subl.
Solubility: Insoluble in water, acids. Soluble in fused alkalis
UN No.: 1479
MDL No.: MFCD00011210
Formel:
O₂Ru Smältpunkt: 1200 °C (subl.) Densitet: 6,97 Storage Temperature: Omgivande |
MDL Number:
MFCD00074857 CAS nummer: 12036-10-1 EINECS: 234-840-6 UN: 1479 ADR: 5.1,II |