Nickelmonoxid, svart (≥76% Ni) (contains Ni2O3)
Leverantör: Thermo Scientific
Fara
Synonyms: Bunsenit, Nickeloxid
87302.30 87302.A1
87302.30EA 2500 SEK
87302.30 87302.A1
Nickelmonoxid, svart (≥76% Ni) (contains Ni2O3)
Nickelmonoxid
CAS No.: 1313-99-1 Molecular Formula: NiO Formula Weight: 74.71 Physical Form: Powder Density: 6.67 Odour: Odourless Merck Reference: 14,6512 MDL No.: MFCD00011145
Formel: NiO
MW: 74,69 g/mol
Smältpunkt: 1984 °C
Densitet: 6,67
Storage Temperature: Omgivande
MDL Number: MFCD00011145
CAS nummer: 1313-99-1
EINECS: 215-215-7
Merck Index: 14,6512