Nickelmonoxid, grön (≥78.5% Ni)
Leverantör: Thermo Scientific
Fara
Synonyms: Bunsenit, Nickeloxid
45094.A1 45094.30
45094.A1EA 7700 SEK
45094.A1 45094.30
Nickelmonoxid, grön (≥78.5% Ni)
Nickelmonoxid
CAS No.: 1313-99-1 Molecular Formula: NiO Formula Weight: 74.71 Physical Form: -400 Mesh Powder Density: 6.67 Odour: Odourless Merck Reference: 14,6512 MDL No.: MFCD00011145
Formel: NiO
MW: 74,69 g/mol
Smältpunkt: 1984 °C
Densitet: 6,67
Storage Temperature: Omgivande
MDL Number: MFCD00011145
CAS nummer: 1313-99-1
EINECS: 215-215-7
Merck Index: 14,6512