Iridium(IV) oxide 99%, pulver
Leverantör: Thermo Scientific
Fara
Synonyms: Iridium dioxide, Iridium(IV) oxide
A17849.03 A17849.06
A17849.03EA 16300 SEK
A17849.03 A17849.06
Iridium(IV) oxide 99%, pulver
Iridium(IV) oxide
Purity: 99% CAS No.: 12030-49-8 Molecular Formula: IrO2 Formula Weight: 224.20 Odour: Odourless UN No.: 1479 MDL No.: MFCD00011065
Formel: IrO₂
MW: 224,22 g/mol
Kokpunkt: –27 °C (1013 hPa)
Smältpunkt: –102 °C
Densitet: 11,7
Storage Temperature: Omgivande
MDL Number: MFCD00011065
CAS nummer: 12030-49-8
EINECS: 234-743-9
UN: 1479
ADR: 5.1,II