Palladium(II) iodide ≥99.998% Pd ≥ 29%, Premion®
Leverantör: Thermo Fisher Scientific
Varning
43084.09 43084.04
43084.09EA 22975 SEK
43084.09 43084.04
Palladium(II) iodide ≥99.998% Pd ≥ 29%, Premion®
Palladium(II) iodide
Purity: 99.998% CAS No.: 7790-38-7 Molecular Formula: PdI2 Formula Weight: 360.23 Physical Form: Powder Density: 6.003 Appearance: Black Melting Point: 350° dec. MDL No.: MFCD00011171
Formel: I₂Pd
MW: 360.23 g/mol
Smältpunkt: 350 °C (dec.)
Densitet: 6.003
Storage Temperature: Omgivande
MDL Number: MFCD00011171
CAS nummer: 7790-38-7
EINECS: 232-203-7