Palladium(II) iodide (≥28% Pd) ≥99.9%
Leverantör: Thermo Fisher Scientific
Varning
11881.04 11881.09
11881.04EA 4625 SEK
11881.04 11881.09
Palladium(II) iodide (≥28% Pd) ≥99.9%
Palladium(II) iodide
Purity: 99.9% CAS No.: 7790-38-7 Molecular Formula: PdI2 Formula Weight: 360.23 Physical Form: Powder Density: 6.003 Appearance: Black Melting Point: 350° dec. MDL No.: MFCD00011171
Formel: I₂Pd
Smältpunkt: 350 °C (dec.)
Densitet: 6,003
Storage Temperature: Omgivande
MDL Number: MFCD00011171
CAS nummer: 7790-38-7
EINECS: 232-203-7