Cobalt(II) fluoride, ultra dry ≥99.99%
Leverantör: Thermo Fisher Scientific
Fara
Ampuled under argon
35740.04 35740.09
35740.04EA 1825 SEK
35740.04 35740.09
Cobalt(II) fluoride, ultra dry ≥99.99%
Cobalt(II) fluoride
Purity: 99.99% CAS No.: 10026-17-2 Molecular Formula: CoF2 Formula Weight: 96.93 Physical Form: Powder, Ampouled under argon Density: 4.46 Melting Point: 1200° Boiling Point: 1400° Merck Reference: 14,2440 UN No.: 2923 MDL No.: MFCD00010941
Formel: CoF₂
Kokpunkt: 1400 °C
Smältpunkt: 1200 °C
Densitet: 4,46
Storage Temperature: Omgivande
MDL Number: MFCD00010941
CAS nummer: 10026-17-2
EINECS: 233-061-9
UN: 2923
ADR: 8,III
Merck Index: 14,2440