Nickeldiklorid, vattenfri ≥99.99%
Leverantör: Thermo Scientific
Fara
Synonyms: Nickel dichloride
43441.14 43441.06
43441.14EA 15300 SEK
43441.14 43441.06
Nickeldiklorid, vattenfri ≥99.99%
Nickeldiklorid
Purity: 99.99% CAS No.: 7718-54-9 Molecular Formula: Cl2Ni Formula Weight: 129.60 Physical Form: Crystalline powder Density: 3.55 Appearance: Yellow Odour: Odourless Melting Point: 1001° Merck Reference: 14,6505 UN No.: 3288 MDL No.: MFCD00011142
Formel: Cl₂Ni
MW: 129,6 g/mol
Smältpunkt: 1001 °C
Densitet: 3,55
Storage Temperature: Omgivande
MDL Number: MFCD00011142
CAS nummer: 7718-54-9
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 14,6505