Nickel(II)bromid 99%
Leverantör: Thermo Scientific
Fara
Synonyms: Nickel dibromide
203360500 203360100 203362500
ACRO203360500EA 1975 SEK
ACRO203360500 ACRO203360100 ACRO203362500
Nickel(II)bromid 99%
Nickel(II)bromid
Formel: Br₂Ni
MW: 218,5 g/mol
Smältpunkt: 963 °C
Densitet: 5,09 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011141
CAS nummer: 13462-88-9
EINECS: 236-665-0
UN: 3077
ADR: 9,III
Merck Index: 12,06585
Order Now
Specifikation testresultat
Appearance Brown Crystalline powder and/or chunks
Titration Argentometric ≥98.5 % (on dry substance)
Water ≤6 % (K.F.)