Iron(II) bromide, vattenfri ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Iron dibromide, Ferrous bromide, Iron(II) bromide
31154.06 31154.14 31154.22
31154.06EA 1225 SEK
31154.06 31154.14 31154.22
Iron(II) bromide, vattenfri ≥98%
Iron(II) bromide
Purity: 98% min CAS No.: 7789-46-0 Molecular Formula: Br2Fe Formula Weight: 215.67 Physical Form: Granular Density: 4.636 Appearance: Green to dark yellow Odour: Odourless Melting Point: 684° Merck Reference: 14,4042 UN No.: 3260 MDL No.: MFCD00016081
Formel: Br₂Fe
MW: 215.66 g/mol
Smältpunkt: 684 °C
Densitet: 4.636
Storage Temperature: Omgivande
MDL Number: MFCD00016081
CAS nummer: 7789-46-0
EINECS: 232-168-8
UN: 3260
ADR: 8 III
Merck Index: 14,4042