Mangandioxid ≥99.997%, Puratronic®
Leverantör: Thermo Scientific
Varning
Synonyms: Brunsten
10805.14 10805.22 10805.06
10805.14EA 5725 SEK
10805.14 10805.22 10805.06
Mangandioxid ≥99.997%, Puratronic®
Mangandioxid
Purity: 99.997% CAS No.: 1313-13-9 Molecular Formula: MnO2 Formula Weight: 86.94 Physical Form: Powder Density: 5.026 Appearance: Black Melting Point: 535° dec. Merck Reference: 14,5730 MDL No.: MFCD00003463
Formel: MnO₂
MW: 86,94 g/mol
Smältpunkt: 535 °C (dec.)
Densitet: 5,026
Storage Temperature: Omgivande
MDL Number: MFCD00003463
CAS nummer: 1313-13-9
EINECS: 215-202-6
Merck Index: 14,5730