Manganese(II) fluoride ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: Manganese(II) difluoride, Manganese difluoride
11527.09 11527.18
11527.09EA 1450 SEK
11527.09 11527.18
Manganese(II) fluoride ≥99%
Manganese(II) fluoride
Purity: 99% CAS No.: 7782-64-1 Molecular Formula: F2Mn Formula Weight: 92.93 Physical Form: Crystalline Powder Density: 3.98 Appearance: Red Melting Point: 856° Solubility: Insoluble in water, alcohol. Soluble in dilute HF, conc. HCl or HNO3 Merck Reference: 14,5729 UN No.: 3288 MDL No.: MFCD00016222
Formel: F₂Mn
MW: 92,93 g/mol
Smältpunkt: 856 °C
Densitet: 3,98
Storage Temperature: Omgivande
MDL Number: MFCD00016222
CAS nummer: 7782-64-1
EINECS: 231-960-0
UN: 3288
ADR: 6.1,III
Merck Index: 14,5729