Manganese(II) bromide, vattenfri 95%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Manganous bromide, Manganese(ii)bromide, Manganese dibromide, Manganese(II) bromide hydrate
A18321.14 A18321.22
A18321.14EA 2000 SEK
A18321.14 A18321.22
Manganese(II) bromide, vattenfri 95%
Manganese(II) bromide
Purity: 95% CAS No.: 13446-03-2 Molecular Formula: MnBr2 Formula Weight: 214.76 Odour: Odourless Melting Point: 698° MDL No.: MFCD00011113
Formel: Br₂Mn
MW: 214.75 g/mol
Smältpunkt: 698
Densitet: 4.38
Storage Temperature: Omgivande
MDL Number: MFCD00011113
CAS nummer: 13446-03-2
EINECS: 236-591-9
Merck Index: 14,5725