Molybden(IV)sulfid ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Molybdenum disulphide
12213.30 12213.A1
12213.30EA 2725 SEK
12213.30 12213.A1
Molybden(IV)sulfid ≥98%
Molybden(IV)sulfid
Purity: 98% CAS No.: 1317-33-5 Molecular Formula: MoS2 Formula Weight: 160.08 Physical Form: Powder Appearance: Black Odour: Odourless Melting Point: 1185° Solubility: Insoluble in water and dilute acids Merck Reference: 14,6236 MDL No.: MFCD00003470
Hydrogenation and isomerization catalyst, dry lubricant
Formel: MoS₂
MW: 160.07 g/mol
Smältpunkt: 1185 °C
Densitet: 4.8 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003470
CAS nummer: 1317-33-5
EINECS: 215-263-9
Merck Index: 12,06318