Chromium(III) fluoride ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: Chromium trifluoride
35741.04 35741.09
35741.04EA 5675 SEK
35741.04 35741.09
Chromium(III) fluoride ≥98%
Chromium(III) fluoride
Purity: 99.98% CAS No.: 7788-97-8 Molecular Formula: CrF3 Formula Weight: 108.99 Physical Form: Powder Density: 3.8 Appearance: Dark green Odour: Odourless Melting Point: 1000° Merck Reference: 14,2223 UN No.: 1756 MDL No.: MFCD00016045
Formel: CrF₃
MW: 108,99 g/mol
Smältpunkt: >1000 °C
Densitet: 3,8
Storage Temperature: Omgivande
MDL Number: MFCD00016045
CAS nummer: 7788-97-8
EINECS: 232-137-9
UN: 1756
ADR: 8,II
Merck Index: 14,2223