Tantal(V)oxid, Patinal®
Leverantör: Merck
Synonyms:
Tantalum(V) oxide, Ditantalum pentaoxide
1.08500.0100
1.08500.1000
1.08500.0100EA
0
SEK
1.08500.0100
1.08500.1000
Tantal(V)oxid, Patinal®
Tantalum(V) oxide
Formel:
O₅Ta₂ MW: 441,89 g/mol Smältpunkt: 1870…1880 °C Densitet: 8,2 g/cm³ (20 °C) |
MDL Number:
MFCD00011254 CAS nummer: 1314-61-0 EINECS: 215-238-2 Merck Index: 14,09056 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...