Tantal(V)oxid ≥99.5%, Spectrographic Grade
Leverantör: Thermo Scientific
Synonyms: Tantalum(V) oxide, Ditantalum pentaoxide
73118.22 73118.14
73118.22EA 7600 SEK
73118.22 73118.14
Tantal(V)oxid ≥99.5%, Spectrographic Grade
Tantalum(V) oxide
Purity: 99.5% CAS No.: 1314-61-0 Molecular Formula: Ta2O5 Formula Weight: 441.89 Physical Form: Powder Density: 8.2 Melting Point: 1800° Merck Reference: 14,9056 MDL No.: MFCD00011254
Formel: O₅Ta₂
MW: 441,89 g/mol
Smältpunkt: 1800 °C
Densitet: 8,2
Storage Temperature: Omgivande
MDL Number: MFCD00011254
CAS nummer: 1314-61-0
EINECS: 215-238-2
Merck Index: 14,9056