Tantalum pentachloride 99.99%
Leverantör: Thermo Scientific
Fara
Synonyms: Tantalum (V) chloride
203910050 203910250 203911000
ACRO203910050EA 940 SEK
ACRO203910050 ACRO203910250 ACRO203911000
Tantalum pentachloride 99.99%
Tantalum pentachloride
Formel: Cl₅Ta
MW: 358,21 g/mol
Kokpunkt: 232…235 °C (1013 hPa)
Smältpunkt: 221 °C
Densitet: 3,68 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011253
CAS nummer: 7721-01-9
EINECS: 231-755-6
UN: 3260
ADR: 8,II
Order Now
Specifikation testresultat
Appearance White to light yellow Crystalline powder or crystals
Total trace metal impurities ≤100 ppm