Titanium(IV) methoxide ≥95%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Titanium tetramethanolate, Titanium (IV) methoxide
77137.14 77137.22 77137.36
77137.14EA 1875 SEK
77137.14 77137.22 77137.36
Titanium(IV) methoxide ≥95%
Titanium(IV) methoxide
Purity: 95% CAS No.: 992-92-7 Molecular Formula: Ti(OCH3)4.(CH3OH)x, x1 Flash Point: 48°(118°F) Physical Form: Granular/ lump Sensitivity: Moisture Sensitive Boiling Point: 243°/52mm UN No.: 3181 MDL No.: MFCD00014883
Formel: C₄H₁₂O₄Ti
MW: 172.02 g/mol
Kokpunkt: 243 °C (52 mmHg)
Smältpunkt: 200…210 °C
Flampunkt: 48 °C (118 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00014883
CAS nummer: 992-92-7
EINECS: 213-596-4
UN: 3181
ADR: 4.1 III