Titanium(IV) isopropoxide 95%
Leverantör: Thermo Scientific
Varning
Synonyms: Titanium tetraisopropanolate, Titanium (IV) isopropoxide
A13703.0E A13703.22 A13703.36
A13703.0EEA 7400 SEK
A13703.0E A13703.22 A13703.36
Titanium(IV) isopropoxide 95%
Titanium(IV) isopropoxide
Purity: 95% CAS No.: 546-68-9 Molecular Formula: C12H28O4Ti Formula Weight: 284.23 Flash Point: 46°(115°F) Refractive Index: 1.4640 Melting Point: 16-20° Boiling Point: 232° Beilstein Reference: 3679474 Merck Reference: 14,9480 UN No.: 2413 MDL No.: MFCD00008871
Formel: C₁₂H₂₈O₄Ti
MW: 284,23 g/mol
Kokpunkt: 232 °C
Smältpunkt: 16…20 °C
Densitet: 0,955
Flampunkt: 46 °C (115 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008871
CAS nummer: 546-68-9
EINECS: 208-909-6
UN: 2413
ADR: 3,III
Merck Index: 14,9480