Titanium(IV) oxide rutile ≥99.9%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Pigment White 6, Rutile, Titanium dioxide rutile
44375.14 44375.22
44375.14EA 2750 SEK
44375.14 44375.22
Titanium(IV) oxide rutile ≥99.9%
Titanium(IV) oxide rutile
Purity: 99.9% CAS No.: 1317-80-2 Molecular Formula: TiO2 Formula Weight: 79.90 Physical Form: 3-6mm Sintered Pieces Density: 4.23 Melting Point: 1830-1850° Boiling Point: 2500-3000° Merck Reference: 14,9472 MDL No.: MFCD00011269
Formel: O₂Ti
MW: 79.88 g/mol
Kokpunkt: 2500…3000 °C
Smältpunkt: 1830…1850 °C
Densitet: 4.23
Storage Temperature: Omgivande
MDL Number: MFCD00011269
CAS nummer: 1317-80-2
EINECS: 215-282-2
Merck Index: 14,9472