Titanium(IV) oxide rutile ≥99.5%
Leverantör: Thermo Fisher Scientific
|
Varning
|
Purity: 99.5% min
CAS No.: 1317-80-2
Molecular Formula: O2Ti
Formula Weight: 79.90
Physical Form: 1.0-2.0 Micron APS Powder
Density: 4.23
Appearance: White
Odour: Odourless
Melting Point: 1830-1850°
Boiling Point: 2500-3000°
Merck Reference: 14,9472
MDL No.: MFCD00011269
Formel:
O₂Ti Kokpunkt: 2500…3000 °C Smältpunkt: 1830…1850 °C Densitet: 4,23 Storage Temperature: Omgivande |
MDL Number:
MFCD00011269 CAS nummer: 1317-80-2 EINECS: 215-282-2 Merck Index: 14,9472 |