Titanium(IV) oxide rutile ≥99.8%
Leverantör: Thermo Fisher Scientific
Varning
42681.30 42681.A1 42681.A7
42681.30EA 2925 SEK
42681.30 42681.A1 42681.A7
Titanium(IV) oxide rutile ≥99.8%
Titanium(IV) oxide rutile
Purity: 99.8% CAS No.: 1317-80-2 Molecular Formula: TiO2, 97% min rutile phase Formula Weight: 79.90 Physical Form: 0.9-1.6 Micron APS Powder, S.A. 3.3-6.1 m2/g Density: 4.23 Melting Point: 1830-1850° Boiling Point: 2500-3000° Merck Reference: 14,9472 MDL No.: MFCD00011269
Formel: O₂Ti
Kokpunkt: 2500…3000 °C
Smältpunkt: 1830…1850 °C
Densitet: 4,23
Storage Temperature: Omgivande
MDL Number: MFCD00011269
CAS nummer: 1317-80-2
EINECS: 215-282-2
Merck Index: 14,9472