Titanium(IV) oxide rutile ≥99.99%
Leverantör: Thermo Fisher Scientific
|
Varning
|
Synonyms:
Pigment White 6, Rutile, Titanium dioxide rutile
14631.09
14631.18
14631.30
14631.09EA
2125
SEK
14631.09
14631.18
14631.30
Titanium(IV) oxide rutile ≥99.99%
Titanium(IV) oxide rutile
Purity: 99.99%
CAS No.: 1317-80-2
Molecular Formula: TiO2
Formula Weight: 79.90
Physical Form: Powder
Density: 4.23
Appearance: White
Odour: Odourless
Melting Point: 1830-1850°
Boiling Point: 2500-3000°
Merck Reference: 14,9472
MDL No.: MFCD00011269
Formel:
O₂Ti MW: 79.88 g/mol Kokpunkt: 2500…3000 °C Smältpunkt: 1830…1850 °C Densitet: 4.23 Storage Temperature: Omgivande |
MDL Number:
MFCD00011269 CAS nummer: 1317-80-2 EINECS: 215-282-2 Merck Index: 14,9472 |