Titan(IV)oxid Reag. Ph. Eur. analytiskt reagens, Supelco®
Leverantör: Merck
Synonyms: Titanic anhydride, Pigment White 6, Rutile, Titanium(IV) oxide, rutile, Titanium dioxide
M-Clarity™ quality level = MQ300
1.00808.1000 1.00808.9025 1.00808.9050
1.00808.1000EA 1550 SEK
1.00808.1000 1.00808.9025 1.00808.9050
Titan(IV)oxid Reag. Ph. Eur. analytiskt reagens, Supelco®
Titanium(IV) oxide
Formel: TiO₂
Kokpunkt: 2900 °C (1013 hPa)
Smältpunkt: 1855 °C
Densitet: 4,2 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011269
CAS nummer: 13463-67-7
EINECS: 236-675-5
Merck Index: 13,09549
Order Now
Specifikation testresultat
Solubility (20 °C) insoluble
Melting Point 1855 °C
Molar Mass 79.87 g/mol
Bulk Density 850 kg/m3
Boiling Point 2900 °C
pH 7 - 8 (100 g/l, H2O, 20 °C) (slurry)