Titanium(IV) oxide anatase catalyst support
Leverantör: Thermo Scientific
Varning
Synonyms: Pigment White 6, Titanium dioxide anatase, Anatase
43828.30 43828.A1 43828.A7
43828.30EA 2175 SEK
43828.30 43828.A1 43828.A7
Titanium(IV) oxide anatase catalyst support
Titanium(IV) oxide anatase
CAS No.: 1317-80-2 Molecular Formula: TiO2 Formula Weight: 79.90 Physical Form: 1/8" Pellets Appearance: White Odour: Odourless MDL No.: MFCD00011269
Formel: TiO₂
MW: 79,88 g/mol
Kokpunkt: 2900 °C (1013 hPa)
Smältpunkt: 1855 °C
Densitet: 4,2 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011269
CAS nummer: 13463-67-7
EINECS: 215-280-1
Merck Index: 13,09549