Titan(IV)oxid predominantly rutile 99.999%
Leverantör: Thermo Fisher Scientific
Synonyms: Titanic anhydride, Pigment White 6, Rutile, Titanium(IV) oxide, rutile, Titanium dioxide
194340050 194340250
ACRO194340050EA 1125 SEK
ACRO194340050 ACRO194340250
Titan(IV)oxid predominantly rutile 99.999%
Titanium(IV) oxide
Formel: TiO₂
MW: 79.88 g/mol
Kokpunkt: 2900 °C (760 mmHg)
Smältpunkt: 1855 °C
Densitet: 4.23 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00011269
CAS nummer: 13463-67-7
EINECS: 236-675-5
Merck Index: 13,09549
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Total trace metal impurities ≤0.001 %