Hafnium dioxide, Patinal®

Leverantör: Merck

1.11529.0100 1.11529.1000
1.11529.0100EA 3650 SEK
1.11529.0100 1.11529.1000
Hafnium dioxide, Patinal®
Hafnium dioxide
Formel: HfO₂
MW: 210,49 g/mol
Kokpunkt: 5400 °C (1013 hPa)
Smältpunkt: 2812 °C
Densitet: 9,68 g/cm³ (20 °C)
MDL Number: MFCD00003565
CAS nummer: 12055-23-1
EINECS: 235-013-2
Merck Index: 14,04727

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR