Hafnium dioxide ≥99.9% (max. 0.5% Zr)
Leverantör: Thermo Scientific
36204.09 36204.18
36204.09EA 2175 SEK
36204.09 36204.18
Hafnium dioxide ≥99.9% (max. 0.5% Zr)
Hafnium dioxide
Purity: 99.9% CAS No.: 12055-23-1 Molecular Formula: HfO2 Formula Weight: 210.49 Physical Form: 3-12mm Sintered Lump Density: 9.68 Melting Point: 2774° Merck Reference: 14,4588 MDL No.: MFCD00003565
Formel: HfO₂
MW: 210,49 g/mol
Kokpunkt: 5400 °C (1013 hPa)
Smältpunkt: 2774 °C
Densitet: 9,68
Storage Temperature: Omgivande
MDL Number: MFCD00003565
CAS nummer: 12055-23-1
EINECS: 235-013-2
Merck Index: 14,4588