Diyttriumtrioxid ≥99.995% (REO, rare earth oxide basis), nanopowder, REacton®
Leverantör: Thermo Scientific
Varning
Synonyms: Yttrium oxide, Yttrium (III) oxide
44048.14 44048.22 44048.36
44048.14EA 3200 SEK
44048.14 44048.22 44048.36
Diyttriumtrioxid ≥99.995% (REO, rare earth oxide basis), nanopowder, REacton®
Diyttriumtrioxid
Purity: 99.995% CAS No.: 1314-36-9 Molecular Formula: Y2O3 Formula Weight: 225.81 Physical Form: 25-50nm APS Powder Density: 5.01 Appearance: White Odour: Odourless Melting Point: 2410° Merck Reference: 14,10107 MDL No.: MFCD00011473
Formel: Y₂O₃
MW: 225,81 g/mol
Kokpunkt: 4300 °C (1013 hPa)
Smältpunkt: 2410 °C
Densitet: 5,01
Storage Temperature: Omgivande
MDL Number: MFCD00011473
CAS nummer: 1314-36-9
EINECS: 215-233-5
Merck Index: 14,10107