Diyttriumtrioxid ≥99.9999% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Scientific
Varning
Synonyms: Yttrium oxide, Yttrium (III) oxide
42864.09 42864.18 42864.30
42864.09EA 3800 SEK
42864.09 42864.18 42864.30
Diyttriumtrioxid ≥99.9999% (REO, rare earth oxide basis), REacton®
Diyttriumtrioxid
Purity: 99.9999% CAS No.: 1314-36-9 Molecular Formula: Y2O3 Formula Weight: 225.81 Physical Form: Powder Density: 5.01 Appearance: White Odour: Odourless Melting Point: 2410° Merck Reference: 14,10107 MDL No.: MFCD00011473
Formel: Y₂O₃
MW: 225,81 g/mol
Kokpunkt: 4300 °C (1013 hPa)
Smältpunkt: 2410 °C
Densitet: 5,01
Storage Temperature: Omgivande
MDL Number: MFCD00011473
CAS nummer: 1314-36-9
EINECS: 215-233-5
Merck Index: 14,10107